Would you like to log in again?
Rapid Response™ Fentanyl Test Strip, BTNX
Supplier: BTNX
FYL-1S48-100
76473-480KT 109.36 USD InStock
76473-480
Rapid Response™ Fentanyl Test Strip, BTNX
Drug Tests
The Rapid Response™ Fentanyl (FYL) Test Strip is a rapid visual immunoassay for the qualitative detection of fentanyl at a cut-off of 200 ng/mL for liquid/powder or 20 ng/mL for urine.
  • Parameter: FYL
  • Calibrator: Fentanyl
  • Cut-off: 200 ng/ml (Liquid/Powder), 20 ng/ml (Urine)
Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no control line).
Delivery information: Includes test strips, product insert.
Caution: This test does not evaluate drug safety or purity. This test is for forensic use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty