Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rapid Response™ Urinalysis Reagent Strips, BTNX

Supplier: BTNX

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Rapid Response™
U2.3-1S25 U2.1-1S100 U7.1-1S100 U5.1-1S100 U10.1-1S100
76473-520KT 32.22 USD InStock
76473-520 76473-518 76473-516 76473-522 76473-512
Rapid Response™ Urinalysis Reagent Strips, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response® Urinalysis reagent strip is a fast, qualitative and semi-quantitative, dip-and-read test for the detection of urine analytes in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases.

  • Sample: Urine
  • Format: Strip
  • Time to result: Within 2 minutes
  • Storage condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Colorimetric analysis

Test results are interpreted by direct comparison to the color blocks printed on the canister label.

Urinalysis reagent strips can include combinations of leukocytes (LEU), nitrite (NIT), urobilinogen (URO), protein (PRO), pH, blood (BLO), specific gravity (SG), ketone (KET), bilirubin (BIL), glucose (GLU).

Delivery information: Includes reagent strips and product insert.

Caution: This test is intended for in vitro diagnostic use only.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty