Would you like to log in again?
TIP ROBOTIC FLTRD ST TECAN 1000UL CS3840
Supplier: AZER SCIENTIFIC INC
Pipet tip, robotic, for automation, Tecan compatible, conductive 1000ul filter tips, sterile
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TIP ROBOTIC FLTRD ST TECAN 1000UL CS3840 76474-060 ES812921000FRS Case of 3,840 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty