Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quidel QuickVue Influenza A+B Test, NDC

Supplier: National Distribution and Contracting
QuickVue
QUI20183
76474-226KT 525.85 USD InStock
76474-226
Quidel QuickVue Influenza A+B Test, NDC
Clinical Diagnostic Supplies Clinical Reagents and Kits
The QuickVue Influenza A+B Test detects and differentiates influenza type A and type B antigens directly from nasal swab and nasopharyngeal swab specimens.

  • PPA - A: 81.5%, B: 80.9%
  • NPA - A: 97.8%, B: 99.1%
  • CLIA complexity: waved

A single sample can be used to run both the QuickVue Influenza A+B Test and QuickVue RSV10 Test.

Processing time: 10 minutes or less

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty