Would you like to log in again?
Justrite® Safety Drum Pumps for 30 and 55 Gallon Drums
Supplier: Justrite
07717 07712
76474-344EA 547.05 USD InStock
76474-344 76474-348
Justrite® Safety Drum Pumps for 30 and 55 Gallon Drums
Pumps Drum/Transfer Pumps
Ideal for small quantity dispensing of liquids.
  • For 30 and 55 gal. drums
  • 8 oz. per stroke
  • 36" reach below lid
  • 22" suction tube length
This manually operated piston pump is FM approved and delivers up to 8 oz. per stroke, ideal for transferring flammable and combustible liquids in a controlled and safe manner. PTFE seals, offering superior chemical resistance and a long life. Included with the pump is a standard 2" NPT threaded bung adapter. The included pick-up tube allows the pump to reach up to 36" below the lid of the drum. Included flash arrestor keeps contents safe from external flame flashbacks. The bonding and grounding clamps come pre-assembled and provide the means to safely ground the receiving container to avoid a static discharge.
FM approved.
Ordering information: One-year warranty.
Delivery information: Includes pump, pick-up tube, 2" NPT bung adapter, grounding wires with clamps, instruction sheet.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty