Would you like to log in again?
Justrite® Safety Drum Pump for 5 Gallon Pails
Supplier: Justrite
07713
76474-346EA 552.56 USD InStock
76474-346
Justrite® Safety Drum Pump for 5 Gallon Pails
Pumps Drum/Transfer Pumps
Ideal for small quantity dispensing of liquids from 5 gallon pails and other containers.
  • Stainless steel
  • For 5 gal. pails
  • 12" reach below lid
This manually operated piston pump is FM approved and delivers up to 3.5 oz. per stroke, ideal for transferring flammable and combustible liquids in a controlled and safe manner. The pump body is constructed of stainless steel with PTFE seals, offering superior chemical resistance and a long life. Included with the pump are five threaded adapters to fit a variety of pail spouts. The pump has a maximum reach of 12" below the lid of the pail. Included flash arrestor keeps contents safe from external flame flashbacks. The bonding and grounding clamps come pre-assembled and provide the means to safely ground the receiving container to avoid a static discharge.
Certifications: FM approved.
Ordering information: One-year warranty.
Delivery information: Includes pump, 5x pail spout adapters, 2x ground wires with clamps, instruction sheet.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty