Would you like to log in again?
FILTER VENT PES GAMMASTAB 0.2UM PK500
Supplier: SARTORIUS CORPORATION EDI
Venting Filter, hydrophobic PES;0.2Um;25mm;gammastab.;500 pc; FLL inlet, MLS outlet
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER VENT PES GAMMASTAB 0.2UM PK500 76474-444 1757HQ Pack of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty