Would you like to log in again?
Honeycomb Grid ESD Curtain, Transforming Technologies
Supplier: Transforming Technologies
ESDC1003C
76474-658EA 498.05 USD InStock
76474-658
Honeycomb Grid ESD Curtain, Transforming Technologies
Surface Protectors Bench Protectors
Anti-Static and ESD curtains are a great way to separate critical workspaces while also attending to the static-sensitive nature of certain products.
  • Ideal barrier wall for sensitive environments
  • Honeycomb conductive scrim grounds static charges
  • Curtain rolls can be easily converted for specific applications
  • Easily cut custom sizes with a razor or knife and replace as needed
  • Made with clear PVC
Transparent PVC ESD curtains are an ideal barrier wall for sensitive environments. Honeycomb grids are installed on both sides of the PVC curtain for safe dissipation of static electricity. These anti-static curtains are critical for applications where any electrical discharge could pose damage to critical components or explosion risk such as electronics, pharmaceutical, medical, printing, or painting. The ESD curtains also have a great draping ability so that they can be used as a protective covering for carts, instruments, or entire walls. The PVC material is very flexible and easy to cut for specific applications. Sold in 4' wide rolls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty