Would you like to log in again?
Single Wire Wrist Band and Coil Cord Sets, Transforming Technologies
Supplier: Transforming Technologies
WB1845 WB1744
76474-732EA 10.69 USD InStock
76474-732 76474-734
Single Wire Wrist Band and Coil Cord Sets, Transforming Technologies
Grounders Wrist Grounders
These versatile wrist strap and coil cord sets are available with a 7- or 10-mm snap. The set is a durable and long-lasting method of grounding personnel.
  • Durable and long-lasting design
  • Comfortable to wear for extended periods
  • Fabric band uses hypoallergenic silver-plated nylon thread
  • One size fit all
This Wrist Band and Coil Cord set is the most versatile and cost-effective band set. The soft, blue woven fabric and 360° curved buckle of the wrist band allows the skin to breathe and makes it very comfortable to wear. The band uses hypoallergenic silver-plated nylon thread for conductivity and has a maximum resistance of 160 ohms. The coil cords 2.5 mm polyurethane coil insulation offers excellent coil memory. A swivel type banana jack increases coil life and prevents cord tangles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty