Would you like to log in again?
Premium Single Wire Wrist Strap and Coil Cord Sets, WB4000 Series, Transforming Technologies
Supplier: Transforming Technologies
WB4037 WB4043 WB4016
76474-754EA 4.91 USD InStock
76474-754 76474-758 76474-756
Premium Single Wire Wrist Strap and Coil Cord Sets, WB4000 Series, Transforming Technologies
Grounders Wrist Grounders
The WB4000 Series Wrist Band and Coil Cord set is a premium band set featuring a machined 4 mm snap post.
  • Soft woven fabric is comfortable to wear for long periods
  • Machined snap provides a more secure connection to the coil cord
  • Cord is durable and has excellent coil memory
The soft wrist band features a 360° conductive contact around the wrist, providing maximum skin contact. The band uses hypoallergenic silver-plated nylon thread for conductivity and has a maximum resistance of 160 ohms. The soft, blue woven fabric of the wrist band allows the skin to breathe and is very comfortable to wear. A shorter, swivel type banana jack increases coil life, prevents cord tangles, and fits most bench grounds.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and ESDA Standard 1.1-2006.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty