Would you like to log in again?
SPECTROMETER MIR-DTC KBR 8300 - 350 CM-1
Supplier: PERKINELMER U.S. LLC
Spectrometer, MIR DTC KBR, Wave Length Range: 8,300 - 350 cm-1, provides the sampling flexibility and performance in mid, near, and far infrared ranges through a single instrument to advance research and new product development in academia, chemicals, polymers, and pharmaceuticals
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPECTROMETER MIR-DTC KBR 8300 - 350 CM-1 76474-974 L1280133 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty