Would you like to log in again?
Epredia™ Premium Microscope Slides in Tropical Packaging, Erie Scientific (Thermo Scientific)
Supplier: Thermo Scientific
Epredia™
3032-002A 3011-002A 3050-002A 3063-002A
76475-892CS 1252.52 USD InStock
76475-892 76475-890 76475-896 76475-894
Epredia™ Premium Microscope Slides in Tropical Packaging, Erie Scientific (Thermo Scientific)
Slides Microscope Slides
Microscope slides are packaged in a sealed tropical barrier bag with desiccant.
  • Manufactured from pure white glass
  • Reduced iron content for low autofluorescence
  • Precleaned for immediate use
  • Non-adhesion style slides
Epredia™ premium microscope slides are supplied in a sealed tropical barrier bag with desiccant to reduce slide surface deterioration and sticking. Manufactured in the USA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty