Would you like to log in again?
Transfer Pipettes, Individual Wrap, Sterile, Cole-Parmer
Supplier: Cole Parmer
06226-42 06226-45 06226-44 06226-43
76476-420BX 68.5 USD InStock
76476-420 76476-422 76476-424 76476-426
Transfer Pipettes, Individual Wrap, Sterile, Cole-Parmer
Pipettes Pasteur/Transfer Pipettes
No more breakage or cross-contamination.
  • Translucent shatterproof LDPE is nontoxic and biologically inert
  • Nonbinding for cells and proteins to prevent sample loss
  • Heat sealable and liquid nitrogen compatible
  • Sterilization method: ethylene oxide
Low-density polyethylene (LDPE) makes these sterile pipettes with built-in bulbs the perfect substitute for breakable glass pipettes, eliminating cross-contamination and cumbersome rubber bulb changes. Plastic reduces the loss of cells and proteins that often occurs with binding that takes place with glass alternatives. Precision manufacturing ensures uniform repeatable drops.
Heat seal pipettes to provide liquid-tight transport and storage. These pipettes are suitable for use with a temperature range of –50 to 90 °C and can be gas sterilized.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty