Would you like to log in again?
Hypersil GOLD™ PEI HILIC HPLC Column
Supplier: Thermo Scientific
Hypersil GOLD™
26505-259070A
76477-176EA 2869.78 USD InStock
76477-176
Hypersil GOLD™ PEI HILIC HPLC Column
Chromatography Columns
Highly polar and hydrophilic analytes that are difficult to analyze with conventional C18 columns can be retained with Thermo Scientific™ Hypersil GOLD™ PEI HILIC LC columns for hydrophilic interaction chromatography.
These columns enable straightforward separations without sample derivatization or ion-pairing reagents. The columns feature excellent peak shape and selectivity and offer improved sensitivity in MS detection.
With particle sizes of 1.9, 3 and 5 µm, Hypersil GOLD PEI-HILIC columns are ideal for applications in drug development as well as in food and environmental analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty