Would you like to log in again?
Hypersil GOLD™ PFP HPLC Column
Supplier: Thermo Scientific
Hypersil GOLD™
25405-259070A
76477-178EA 2520.36 USD InStock
76477-178
Hypersil GOLD™ PFP HPLC Column
Chromatography Columns
Analyze difficult-to-solve mixtures of halogenated compounds and non-halogenated polar compounds with Thermo Scientific™ Hypersil GOLD™ PFP (pentafluorophenyl) HPLC columns.
These columns offer an alternative selectivity to C18 columns in reversed phase applications with excellent peak shape and sensitivity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty