Would you like to log in again?
Hypersil™ Chiral-IZT HPLC Column
Supplier: Thermo Scientific
Hypersil™
42405-054630
76477-184EA 1821.94 USD InStock
76477-184
Hypersil™ Chiral-IZT HPLC Column
Chromatography Columns
Separate a wide range of enantiomers with Thermo Scientific™ Hypersil™ Chiral HPLC columns, designed for normal-phase separations of neutral, acidic, and basic racemates.
These chiral columns help meet your purification needs by resolving racemic mixtures and potential contaminants so you can be confident in the purity of your target molecule.
To maximize screening efficiency, these columns can be ordered in kits containing one column in each of our chiral phases. All columns are available in analytical and preparative scaled formats.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty