Would you like to log in again?
CAP SURGEON SMS NON-WOVEN BLUE CS100
Supplier: DUKAL CORPORATION
Surgeons Cap, Non-Woven, Made of multi-layer material which offers enhanced protection, soft and absorbent, Built in sweat band helps to control sweat and moisture inside the cap, Color: Blue, Material: SMS, Not made with natural rubber latex
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP SURGEON SMS NON-WOVEN BLUE CS100 76478-460 312 Case of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty