Would you like to log in again?
COVERALL PYROLON CRFR WITH HOOD L CS6
Supplier: CHEMLAND-LAKELAND INDUS
Coverall, Pyrolon CRFR, with Hood, Storm flap over, zipper, Elastic face, Elastic wrists and ankles, bar contaminating flammables like paint, oil andgrease, hazardous liquids and contaminants, and dry particulates from penetrating to inner Primary Protective Garments, Size: 2x
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALL PYROLON CRFR WITH HOOD L CS6 76478-534 51130-LG Case of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty