Would you like to log in again?
SUIT ENCAPSULATED BACK/REAR ENTRY 3X CS3
Supplier: CHEMLAND-LAKELAND INDUS
Suit, Encapsulated, Expanded Back/Rear Entry, Heat Sealed, Yellow, Materials: Spun bonded polypropylene & Proprietary barrier film, 48in zipper, Double storm flap, 20 mil Vinyl face shield, Elastic wrists, 2 exhaust ports with shroud, Air tube inlet, Attached sock boots with boot flap, Size:32x
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SUIT ENCAPSULATED BACK/REAR ENTRY 3X CS3 76478-550 C4T450Y-3X Case of 3 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty