Would you like to log in again?
LAB COAT COLLAR ELASTIC WRIST 3X CS30
Supplier: CHEMLAND-LAKELAND INDUS
Lab coat, Elastic wrist, Snap closure, Collar, Materials: 55 gram SBPP with Laminated Microporous Film, No pockets, Long sleeve, Lightweight, Features high MVTR (Moisture Vapor Transmission Rate), Protection from dry particulates and light liquid splash, Size: 3x
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAB COAT COLLAR ELASTIC WRIST 3X CS30 76478-564 CTL112-3X Case of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty