Would you like to log in again?
LABCOAT SNAP CLOSURE LONG SLEEVE 2X CS30
Supplier: CHEMLAND-LAKELAND INDUS
Lab coat, Snap closure, Snap closure, No pockets, Long sleeves, Materials: 55 gram SBPP with Laminated Microporous Film, No pockets, Long sleeve, Lightweight, Features high MVTR (Moisture Vapor Transmission Rate), Protection from dry particulates and light liquid splash, Size: XL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LABCOAT SNAP CLOSURE LONG SLEEVE 2X CS30 76478-572 CTL140-2X Case of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty