Would you like to log in again?
COVERALL COOL SUIT WITH COLLAR L CS25
Supplier: CHEMLAND-LAKELAND INDUS
Coverall, Suit Coverall, Features: Collar, Open wrists & ankles, Breathable back panel, Materials: 55 gram SBPP with Laminated Microporous Film, Zipper closure, Front and sides material & Storm flap over zipper provides protection from dirt, liquids and light chemical splash, Size: L
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALL COOL SUIT WITH COLLAR L CS25 76478-582 COL412-LG Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty