Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SIMPORT® Transport Tubes, Self-Standing, Simport Scientific

Supplier: Simport

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Simport®
T552-10ATTP T552-12ATTP T552-7ATTP T552-7ATTP T552-12ATTPS T552-30ATTP T552-10ATTP T552-5ATTP T552-12ATTP T552-30ATTP T552-5ATTP
76413-650CS 127.79 USD InStock
76413-650 CA76413-650 76413-654 CA76413-654 76413-656 CA76413-656 76478-618 76413-646 76413-648 CA76413-646 CA76413-648
SIMPORT® Transport Tubes, Self-Standing, Simport Scientific
Tubes Test Tubes
Tubes made of non-toxic polypropylene, caps made of polyethylene. Designed for storage and transportation of biological material.

  • Good chemical resistance
  • Good temperature resistance
  • All graduated tubes are in 5 ml increments

Designed for storage and transportation of biological material. Tubes provide strength and clarity and exhibit some unique design features. Five sizes are available from 5 to 30 ml. They have external threads to provide a smooth and uniform inner surface.

A perfect leakproof seal is obtained by the use of a specially designed flexible sealing lip inside the polyethylene closures. Caps also feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn.

Certifications: 95 kPa tested.

Ordering information: Screw caps have to be ordered separately.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty