Would you like to log in again?
WIPE NONWOVEN DISP WHT 13.75X12IN CS3000
Supplier: MEDLINE INDUSTRIES
Wipe, Disposable, Nonwoven, White, 13.75x12in, Excellent strength and absorbency with low-lint properties, Ideal material for heavy-duty cleaning, wiping, degreasing and surface prep, Material: Polypropylene (PP)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WIPE NONWOVEN DISP WHT 13.75X12IN CS3000 76478-824 MDT217DC30 Case of 3,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty