Would you like to log in again?
HyFlex® 11-754 Light-Duty Industrial Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyFlex®
11754090 11754080 11754120 11754120 11754110 11754070 11754100 11754090 11754070 11754060 11754100 11754080 11754060 11754110
76478-884CS 1555.02 USD InStock
76478-884 CA76478-882 76478-886 CA76478-884 76478-890 76478-880 CA76478-890 CA76478-880 76478-882 CA76478-878 CA76478-886 76478-888 76478-878 CA76478-888
HyFlex® 11-754 Light-Duty Industrial Gloves, Ansell
Gloves
Lightest weight ANSI A4/ ISO Cut D glove featuring INTERCEPT™ Technology, these ultra-thin breathable gloves offer high cut resistance, while maintaining enhanced flexibility and tactility.
  • Ultra-thin ANSI Cut level A4 / ISO Cut level D gloves
  • 3x more cut resistance for unmatched protection
  • Touchscreen compatible for enhanced productivity
  • Ultra-lightweight design provides dexterity for handling and assembly of sharp parts
Ultra-thin breathable gloves are touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks. Designed to fit the natural shape of the hand to help reduce hand fatigue. Clean and skin friendly, HyFlex® 11-754 gloves are dermatologically approved by Dermatest®.
Certifications: REACH compliant. EN 388 3X41D. ANSI 4 ABR. ANSI A4 CUT.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty