Would you like to log in again?
ABS® Countertop Pharmacy Freezers, Freestanding, Premier Series, Horizon Scientific
Supplier: Horizon Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH-ABT-HC-UCFS-0120A-CAD PH-ABT-HC-UCFS-0120
76478-924EA 3694.52 USD InStock
76478-924 76478-922
ABS® Countertop Pharmacy Freezers, Freestanding, Premier Series, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
The Premier Series countertop freezers deliver quality pharmaceutical results in compact form.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port (³/₈")
  • HFC-free refrigerant (R600a)
The Premier Series countertop freezers offer high-quality cooling in the convenience of a compact space. These units offer premium features for pharmacy storage including a series of temperature alarms to keep stored products at maximum effectiveness. Units come equipped with a temperature monitoring device (TMD), adjustable shelves, and probe access. These freezers run on HFC-free refrigerant for economic efficiency and environmental safety.
The CAD unit is ADA compliant. Units include a two year parts and labor warranty.
Certifications: UL/C-UL listed.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty