Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Syringe Filters
Supplier: VWR
VWR®
SFPES030045SAV SFPT025022NBAV SFCA025045NAV SFPT013022NBAV SFCA025045SAV SFNY030022SAV SFPV013022SAV SFPT013045NBAV SFPT025022NLAV SFPES025022SAV SFPT013045NLAV SFGF025100NAV SFPES025045NAV SFNY013022NAV SFPV030045SAV SFPV013022NAV SFRC030022NAV SFPES030045NAV SFPES025045SAV SFPT013022NLAV SFGF030100NAV SFPV030045NAV SFNY030045SAV SFRC013045NAV SFCA025022SAV SFCA025022NAV SFGF013100NAV SFRC013022NAV SFPES030022SAV SFNY013045NAV SFPT025045NLAV SFPES030022NAV SFPV013045NAV SFPES025022NAV SFPV013045SAV SFPV030022SAV SFPT025045NBAV SFNY025045NAV SFNY025022NAV SFRC030045NAV SFPV030022NAV
76478-982CS 3037.59 USD InStock
76478-982 76478-984 76478-980 76479-008 76479-006 76479-028 76479-012 76479-056 76479-034 76479-010 76479-054 76479-032 76479-038 76479-016 76479-036 76479-058 76479-014 76479-030 76479-052 76478-998 76479-050 76478-994 76478-996 76478-990 76478-992 76479-018 76479-000 76479-022 76479-044 76479-042 76479-020 76479-048 76479-026 76479-004 76479-024 76479-002 76479-046 76479-040 76478-986 76478-988 76479-060
VWR® Syringe Filters
Filters Syringe Filters
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
  • Sterile blister packed filters are Ideal for sterile filtration
  • Nonsterile syringe filters are excellent for HPLC applications
  • Biosafe according to Class VI plastics tests
  • Available with Nylon, PES, PTFE, PVDF, CA, RC membrane, and GF (Glass fiber)
  • Available with hydrophilic or hydrophobic PTFE
  • Each filter has a female Luer-lock inlet and a male slip Luer outlet
Nylon and PTFE membranes are ideal for sample preparation and small volume chemical filtration.
Syringe filters with CA (cellulose acetate) or PES (polyether sulfone) membranes are ideal for sterile filtration of buffers, tissue culture media or additives, and other biologicals. The PES membrane offers high flow rates.
RC and PTFE hydrophilic type offer the best chemical resistance for both aqueous solutions and solvents.
Certifications: Manufactured under ISO9001:2015 quality management.
Caution: Hydrophobic PTFE is not intended for use with aqueous solutions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty