Would you like to log in again?
KIT SHAKER 25CUFT/CELL ROLLER-READY
Supplier: CARON PRODUCTS & SERVICE
Shaker/Cell Roller-Ready kit to replace reinforced shelving on a 25 cu. Ft. Shaker Ready CO2 Incubator, Shaker/Cell Roller-Ready kit is optimized for suspension cell culture using a shaker and 3-tier cell roller. Includes shaker support shelf and roller ramp/support.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT SHAKER 25CUFT/CELL ROLLER-READY 76479-066 CELL303 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty