Would you like to log in again?
COIL E-COATED FOR 22CF 7300 CHAMBERS
Supplier: CARON PRODUCTS & SERVICE
Evaporator coil, Electrostatically coated R290 for 22 CF 73xx Series Chambers. For use with corrosive environment applications. Factory installed at time of order.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COIL E-COATED FOR 22CF 7300 CHAMBERS 76479-086 COAT320 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty