Would you like to log in again?
HUMIDITY CONTROL W/DEHUM 7300/7310 22FT3
Supplier: CARON PRODUCTS & SERVICE
Humidity control, with Dehum, from ambient to 85% RH for 7300 & 7310 22 ft3 series. Includes humidity display/readout. Actively humidifies with gVapor and dehumidifies with evaporator. Factory installed at the time of order.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HUMIDITY CONTROL W/DEHUM 7300/7310 22FT3 76479-106 HUMD316 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty