Would you like to log in again?
Linearity FD iPTH for Siemens Centaur, AUDIT MircoControls
Supplier: AUDIT Microcontrols
K813M-5
76479-168EA 457.34 USD InStock
76479-168
Linearity FD iPTH for Siemens Centaur, AUDIT MircoControls
Media for Microbiology Dehydrated Media for Microbiology
Intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for iPTH.
The Linearity FD iPTH for Siemens Centaur is a five level linearity set. Each level contains the following analytes: iPTH. These five levels demonstrate a linear relationship to each other for their respective analytes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty