Would you like to log in again?
Histosette® II Tissue Processing and Biopsy Cassettes, Simport
Supplier: Simport
Histosette® II
M385-5 M386-3 M386-9 M385-3 M386-5 M385-8 M386-6 M386-7 M385-11 M386-2 M386-4 M385-7 M386-10 M385-12 M385-6 M385-10 M385-9 M386-12 M385-4 M386-11 M386-8 M385-2
76479-228CS 128.72 USD InStock
76479-228 76479-206 76479-218 76479-226 76479-204 76479-216 76479-238 76479-208 76479-242 76479-220 76479-210 76479-232 76479-240 76479-230 76479-224 76479-246 76479-236 76479-214 76479-222 76479-244 76479-234 76479-212
Histosette® II Tissue Processing and Biopsy Cassettes, Simport
Embedding Cassettes Biopsy Processing Cassettes
These patented cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.
  • Made of acetal
  • 45° Anterior writing area
  • Covers removed and packaged separately
  • Secure locking device safely holds specimen during processing
  • Ball and joint front hinge with back closure
  • Compatible with Primera© cassette printers
Most convenient for cassette labeling instruments since covers are already removed from cassettes and are packaged separately in the case. The efficient flow-through slots of these cassettes maximize fluid exchange and ensure proper drainage.
The anterior writing area is slanted at a 45° angle to make the cassette more suitable to be used with certain types of cassette labeling instruments. Each case contains 2 dispenser boxes of 500 cassettes and 1 dispenser box of 1000 covers.
Ordering information: Available in 11 non-cytotoxic, non-metallic colors. Each case contains four dispenser boxes of 250 preassembled cassettes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty