Would you like to log in again?
Unisette™ Tissue Processing and Biopsy Cassettes for Primera© Printers, Simport
Supplier: Simport
UNISETTE™
M405-2 M406-10 M405-5 M406-7 M405-3 M406-12 M406-5 M406-9 M405-4 M405-8 M405-7 M406-2 M406-4 M405-9 M405-11 M405-6 M405-10 M406-3 M405-12 M406-11 M406-8 M406-6
76479-248CS 173.02 USD InStock
76479-248 76479-286 76479-264 76479-254 76479-276 76479-284 76479-262 76479-252 76479-274 76479-268 76479-258 76479-266 76479-288 76479-256 76479-278 76479-290 76479-260 76479-282 76479-250 76479-272 76479-280 76479-270
Unisette™ Tissue Processing and Biopsy Cassettes for Primera© Printers, Simport
Embedding Cassettes Biopsy Processing Cassettes
These cassettes keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage.
  • Made of acetal
  • 35° anterior writing area
  • Large labeling area for easy identification
  • One-piece integral lid that can be opened and closed up to three times without breakage
  • Secure locking system without danger of specimen loss
  • Compatible with Primera© Cassette printers
Molded from acetal, the one-piece integral lid eliminates the need for separate steel lids.The snap latch and hinge-lock design of the UNISETTE™ prevent early separation of base and lid and allows for one-hand operation. Each case contains three dispenser boxes of 500 cassettes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty