Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
MagSnap® 360™ Dual-Wire Coil Cord, Desco
Supplier: Desco Industries
19906
76479-386EA 28.15 USD InStock
76479-386
MagSnap® 360™ Dual-Wire Coil Cord, Desco
Grounders Grounding Wires
The MagSnap® 360™ dual wire coil cord offers a 360° rotation and magnetic connection between the wristband and coil cord.
  • Magnetic connection between wrist band and coil cord
  • 360° rotation between wrist band and cord promotes comfort, freedom of motion, and reduces strain on operator’s wrist
  • Strain relief tests over 1 million cycles; 62 times the minimum requirement per ESD S1.1
  • Right angle phono plug connector with 3.5 mm works with standard dual wire monitors and grounding jacks
  • Relaxed retraction coil cord features a soft pull that does not tug on the wearer’s arm
Magnetic connection between wristband and coil cord creates a superior electrical connection, and eliminates intermittent failures in traditional snap connections using springs, extends the lifespan of the wrist strap, and maintains a 1 to 5 pound perpendicular breakaway force to prevent accidental disconnects. Wristband creates a redundant path-to-ground for an operator with resistance <3.5×10⁷ ohms per the required limits of ANSI/ESD S20.20 for Method 1. If one conductor is damaged, the operator will maintain the path-to-ground through the other conductor. Ideal for use with low voltage style monitors. Date coded so the lifespan of the product may be tracked to ensure quality control.
Certifications: UL listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty