For monitoring and validating the sterilization process.
- Wide range of bioindicators for:
- Steam
- Ethylene oxide
- Dry heat
- Hydrogen peroxide
- Irradiation
Available in ampoule, self-contained, strip, suspension and coupon formats in several materials such as glass, aluminum, poliflex, steel and PVC.
All biological indicators have a certificate reporting D and Z values, survival time, kill time, population and purity.
Certifications: Conform in accordance with ISO 11138 and EN 866 regulations.