Would you like to log in again?
Contam Swabs, Liofilchem, Inc.
Supplier: Liofilchem
86102 86101 86104 86100 86103 86105
76479-550PK 144.85 USD InStock
76479-550 76479-560 76479-554 76479-552 76479-558 76479-556
Contam Swabs, Liofilchem, Inc.
Monitoring Systems Hygiene Monitoring Systems
Contam swabs for microbial monitoring of surfaces.
  • Easy to use
  • Clear color changes
  • No additional reagents
  • Results in 18 to 24 hours
  • 10 CFU/ml detection limit
Contam swabs available in contamination, E.coli-coliforms, listeria, salmonella, yeasts & moulds, and MRSA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty