Would you like to log in again?
VWR TUBING 70A PVC PVC 1X1.25 50FT
Supplier: VWR International
VWR Tubing, 70A, Material: PVC, 1X1.25 50Ft, phthalate, DEHP, BPA free. It ensures safe transfer of fluids, gases, air in food, beverage, medical device, health care markets, NSF-51 listed, meets USP Class VI requirements, is made from non-toxic ingredients conforming to FDA, 3A standards.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR TUBING 70A PVC PVC 1X1.25 50FT 76480-170 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty