Would you like to log in again?
LTS® Filtered Pipette Tips, Rainin® LTS® Compatible
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LTS®
ES80352 ES80358 ES80357 ES80359 ES80355
76480-430PK 216.96 USD InStock
76480-430 76480-432 76480-434 76480-436 76480-428
LTS® Filtered Pipette Tips, Rainin® LTS® Compatible
Pipette Tips
Our sterile, 20, 200 and 1000 µl LTS® pippete filter tips are manufactured from super clear high quality polypropylene. The surfaces of the tips are produced through a special process. This process makes the tip inner surface become super hydrophobic, thus significantly reduces sample loss and provides substantially higher reproducibility when working with critical media.
  • LTS® compatible
  • Low retention for optimal precision and minimal sample loss
  • Barrier filter to prevent aerosol contamination
  • Certified DNase-/RNase-/pyrogen-/endotoxin-free
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty