Would you like to log in again?
ExacTip™ Universal Pipette Tips
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ES80652 ES80759 ES80659 ES80755 ES80655 ES80650
76474-100CS 133.54 USD InStock
76474-100 76480-438 76474-104 76474-102 76474-108 76474-106
ExacTip™ Universal Pipette Tips
Pipette Tips
Azer Scientific’s ExacTip™ pipette tip line is the ideal choice for a universal pipette tip or universal filtered pipette tip. The surface and material of the filter tips utilize special processes and polypropylene resins to ensure the low retention qualities for ensuring they are one of the most accurate pipette tips in the market.
  • Manufactured in a Class 100,000 cleanroom and sterilized under ISO 11137 guidelines
  • Tips have low DNA and protein binding treatment
  • DNase, RNase, and Pyrogren free
  • Sterile individually wrapped racks of 96 tips
  • Contain small micron hydrophobic barrier filters to prevent contaminating aerosols from entering your pipette
These filtered pipette tips are manufactured under stringent ISO 13485:2016 and ISO 9001:2015 quality control guidelines in state-of-the-art Class 100,000 cleanroom environments.
Universal filtered pipette tips give you the flexibility of pipette choice, eliminating the need to purchase expensive brand-specific tips for your pipettor. The manufacturing and sterilization processes allow for the pipette tips to be used for many laboratory applications including molecular biology, cell culture, PCR, toxicology and much more.
Certifications: These filter tips are certified to be DNsae-/RNase-free, pyrogen-free, and PCR inhibitor-free.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty