Would you like to log in again?
FREEZEBOX PACKAGE EXTENDED HEIGHT CF BL
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
FreezeBox Extended Height CL, Blue, Iincludes 1 each of FreezeBox Extended Height, CL Cooling Core -0.5C­ - 4C, CME Extender and CM-03 Tube Module for 30 x 5mL tubes 60mm, 152 x 170 x 123mm, Uniform, low temperature protection for consistent sample results, typically +/-0.1 C or taller
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FREEZEBOX PACKAGE EXTENDED HEIGHT CF BL 76480-466 FZB-CL-L3 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty