Would you like to log in again?
Caliber Fluor™ 580, AAT Bioquest
Supplier: AAT Bioquest
95
76480-540EA 361.89 USD InStock
76480-540
Caliber Fluor™ 580, AAT Bioquest
Assays Cellular Assays
Caliber Fluor™ 580 is an extremely stable fluorophore that has its absorption and fluorescence independent of pH change from pH 3 to 10.
  • pH-Independent Fluorescence
  • High Stability
Caution: For Research Use Only
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty