Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
ICG amine ≥95%
Supplier: AAT BIOQUEST INC
188
76480-712EA 361.89 USD InStock
76480-712
ICG amine ≥95%
ICG amine
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
  • Dependable in vivo imaging applications
  • Infrared absorption wavelength
  • Ideal for bioconjugation
Caution: For Research Use Only
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty