Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-TAMRA Maleimide [Tetramethylrhodamine-5-maleimide] ≥90%

Supplier: AAT BIOQUEST INC
Warning

421
76480-894EA 240.68 USD InStock
76480-894
5-TAMRA Maleimide [Tetramethylrhodamine-5-maleimide] ≥90%
5-TAMRA Maleimide [Tetramethylrhodamine-5-maleimide]
Tetramethylrhodamine-5-maleimide is the purified single isomer of tetramethylrhodamine-5 (and 6)-maleimide mixed isomers.


MW: 481.51 g/mol
CAS Number: 154480-30-5
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty