Would you like to log in again?
Texas Red® Maleimide, Single Isomer, AAT Bioquest
Supplier: AAT Bioquest
483
76480-924EA 118.54 USD InStock
76480-924
Texas Red® Maleimide, Single Isomer, AAT Bioquest
Assays Cellular Assays
Texas Red® maleimide is an excellent thiol-reactive fluorescent reagent for labeling thiol-containing biological molecules (such as peptides, proteins and thiol-modified oligonucleotides).
  • Identical spectra to Texas Red
  • High Stability
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty