Would you like to log in again?
Texas Red® Azide, Single Isomer, AAT Bioquest
Supplier: AAT Bioquest
484
76480-926EA 180.84 USD InStock
76480-926
Texas Red® Azide, Single Isomer, AAT Bioquest
Assays Cellular Assays
Texas Red® azide is an excellent alkyne-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Identical spectra to Texas Red
  • High Stability
Caution: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty