Would you like to log in again?
ReadiLink™ Protein Conjugation Stop Buffer (1 ml), AAT Bioquest
Supplier: AAT Bioquest
5400
76481-784EA 93.54 USD InStock
76481-784
ReadiLink™ Protein Conjugation Stop Buffer (1 ml), AAT Bioquest
Protein Linking and Labeling Systems
ReadiLink™ Protein Conjugation Stop Buffer is used to stop the protein conjugation reactions that use succinimidyl ester, sulfonyl chloride or isothiocyanate.
  • Usable at room temperature
  • Stops all major protein conjugation reactions
  • Simple protocol
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty