Would you like to log in again?
Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence* , AAT Bioquest
Supplier: AAT Bioquest
5523
76481-798EA 240.68 USD InStock
76481-798
Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence* , AAT Bioquest
Assays Enzyme Assays
A variety of crosslinking reagents with a maleimide group are widely used for crosslinking proteins to proteins or proteins to other biomolecules.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty