Would you like to log in again?
Amplite® Rapid Fluorimetric Glutathione GSH/GSSG Ratio Assay Kit *Green Fluorescence* , AAT Bioquest
Supplier: AAT Bioquest
10060
76481-864EA 361.89 USD InStock
76481-864
Amplite® Rapid Fluorimetric Glutathione GSH/GSSG Ratio Assay Kit *Green Fluorescence* , AAT Bioquest
Assays Enzyme Assays
When cells are exposed to increased levels of oxidative stress, GSSG will accumulate and the ratio of GSH to GSSG will decrease.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty