Would you like to log in again?
Amplite® Blue Peroxidase Substrate, AAT Bioquest
Supplier: AAT Bioquest
11005
76481-878EA 180.84 USD InStock
76481-878
Amplite® Blue Peroxidase Substrate, AAT Bioquest
Assays Enzyme Assays
A variety of substrates are available for detecting peroxidase activity in ELISA assays. Colorimetric HRP substrates (e.g., TMB, OPD and ABTS) have been widely used for years. Each of these substrates varies greatly with respect to its performance characteristics such as detection sensitivity, working range and attainable signal-to-noise ratios. Amplite® Blue peroxidase substrate is a soluble fluorogenic substrate for detection of peroxidase activity. Amplite® Blue allows rapid HRP detection assays to be performed with greater sensitivity than the colorimetric HRP substrates (such as TMB and OPD). The fluorescent product of Amplite® Blue does not photobleach. Amplite® Blue exhibits a flat baseline in assays, which facilitates low-level detection sensitivity and allows for high signal-to-noise ratios.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty