Would you like to log in again?
Signal Guard™ HRP Conjugate Stabilizer, AAT Bioquest
Supplier: AAT Bioquest
11010
76481-882EA 118.54 USD InStock
76481-882
Signal Guard™ HRP Conjugate Stabilizer, AAT Bioquest
Assays Immunological Assays
The major advantage of a liquid HRP-conjugate is that it eliminates the chance for human error when dissolving or diluting the lyophilized or concentrated conjugate.
  • Permits liquid-hrp conjugates to be stored at 4°c for 24 months or RT for 6 months
  • Can create stock solutions or ready-to-use reagents
Caution: For research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty